ADVH49B828A7 AChemBlock

4-ethynyl-2-methyl-cyclohexanamine hydrochloride

  • Molecular Weight : 173.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-ethynyl-2-methyl-cyclohexanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85381
Availability details for
Quantity
  • Availability
  • Add to Cart