ADVH49B828A9 AChemBlock

4-chloropyrrolo[2,1-f][1,2,4]triazine-6-carboxylic acid

  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloropyrrolo-2-1-f-1-2-4-triazine-6-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85382
Availability details for
Quantity
  • Availability
  • Add to Cart