ADVH49B828AF AChemBlock

4-Carboxy-2-fluorophenylboronic acid ethylene glycol ester

  • Molecular Weight : 227.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-carboxy-2-fluorophenylboronic-acid-ethylene-glycol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70443
Availability details for
Quantity
  • Availability
  • Add to Cart