ADVH49B828B0 AChemBlock

4-Butoxy-3,5-dichlorophenylboronic acid pinacol ester

  • Molecular Weight : 363.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/4-butoxy-3-5-dichlorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70674
Availability details for
Quantity
  • Availability
  • Add to Cart