ADVH49B828B7 AChemBlock

4-Bromo-2-(trifluoromethoxy)phenylboronic acid pinacol ester

  • Molecular Weight : 384.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-trifluoromethoxy-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70806
Availability details for
Quantity
  • Availability
  • Add to Cart