ADVH49B828C0 AChemBlock

4,5-Dimethylpyridine-3-boronic acid pinacol ester

  • Molecular Weight : 251.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-5-dimethylpyridine-3-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70857
Availability details for
Quantity
  • Availability
  • Add to Cart