ADVH49B828C2 AChemBlock

4,5-Difluoro-2-isopropoxyphenylboronic acid pinacol ester

  • Molecular Weight : 316.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-5-difluoro-2-isopropoxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70858
Availability details for
Quantity
  • Availability
  • Add to Cart