ADVH49B828C9 AChemBlock

4-(4-Bromophenoxy)phenylboronic acid pinacol ester

  • Molecular Weight : 393.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-bromophenoxy-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70601
Availability details for
Quantity
  • Availability
  • Add to Cart