ADVH49B828D0 AChemBlock

3-Fluoro-4-methyl-2-(methylthio)phenylboronic acid pinacol ester

  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-4-methyl-2-methylthio-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70643
Availability details for
Quantity
  • Availability
  • Add to Cart