ADVH49B828D1 AChemBlock

3-Fluoro-2-formyl-5-methylphenylboronic acid pinacol ester

  • Molecular Weight : 282.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-2-formyl-5-methylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70768
Availability details for
Quantity
  • Availability
  • Add to Cart