ADVH49B828D2 AChemBlock

3-Fluoro-2-formyl-4-methoxyphenylboronic acid pinacol ester

  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-2-formyl-4-methoxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70678
Availability details for
Quantity
  • Availability
  • Add to Cart