ADVH49B828E2 AChemBlock

3-Acetyl-4-fluorophenylboronic acid pinacol ester

  • Molecular Weight : 282.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-acetyl-4-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70776
Availability details for
Quantity
  • Availability
  • Add to Cart