ADVH49B828E6 AChemBlock

3,6-Dichloro-2-fluorophenylboronic acid pinacol ester

  • Molecular Weight : 308.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-6-dichloro-2-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70868
Availability details for
Quantity
  • Availability
  • Add to Cart