ADVH49B828E7 AChemBlock

3,5-Dichloro-2-methylphenylboronic acid pinacol ester

  • Molecular Weight : 305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/3-5-dichloro-2-methylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70681
Availability details for
Quantity
  • Availability
  • Add to Cart