ADVH49B828E8 AChemBlock

3,4-Difluoro-5-formylphenylboronic acid pinacol ester

  • Molecular Weight : 286.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-difluoro-5-formylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70772
Availability details for
Quantity
  • Availability
  • Add to Cart