ADVH49B828F0 AChemBlock

methyl 3-amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylate

  • Molecular Weight : 278.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84929
Availability details for
Quantity
  • Availability
  • Add to Cart