ADVH49B828F1 AChemBlock

methyl 3-{[(tert-butoxy)carbonyl]amino}-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylate

  • Molecular Weight : 378.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-tert-butoxy-carbonyl-amino-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84930
Availability details for
Quantity
  • Availability
  • Add to Cart