ADVH49B828FB AChemBlock

methyl 2-aminocyclobutane-1-carboxylate; trifluoroacetic acid

  • Molecular Weight : 243.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-aminocyclobutane-1-carboxylate-trifluoroacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84939
Availability details for
Quantity
  • Availability
  • Add to Cart