ADVH49B828FD AChemBlock

tert-butyl 4-[[(3S)-tetrahydrofuran-3-yl]methyl]piperazine-1-carboxylate

  • Molecular Weight : 270.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-3s-tetrahydrofuran-3-yl-methyl-piperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84620
Availability details for
Quantity
  • Availability
  • Add to Cart