ADVH49B828FE AChemBlock

tert-butyl 4-[(1S)-1-amino-2,2,2-trifluoro-ethyl]piperidine-1-carboxylate

  • Molecular Weight : 282.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-1s-1-amino-2-2-2-trifluoro-ethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84621
Availability details for
Quantity
  • Availability
  • Add to Cart