ADVH49B82903 AChemBlock

methyl 2-[(pyrrolidin-2-yl)formamido]acetate

  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-pyrrolidin-2-yl-formamido-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84947
Availability details for
Quantity
  • Availability
  • Add to Cart