ADVH49B82907 AChemBlock

methyl 2-(tert-butoxycarbonylamino)-6-azaspiro[3.4]octane-3-carboxylate

  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-tert-butoxycarbonylamino-6-azaspiro-3-4-octane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84950
Availability details for
Quantity
  • Availability
  • Add to Cart