ADVH49B8290B AChemBlock

methyl 2-(6-bromo-8-fluoro-3-quinolyl)acetate

  • Molecular Weight : 298.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-6-bromo-8-fluoro-3-quinolyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84954
Availability details for
Quantity
  • Availability
  • Add to Cart