ADVH49B8290C AChemBlock

tert-butyl (6S)-6-(bromomethyl)-5-azaspiro[2.4]heptane-5-carboxylate

  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6s-6-bromomethyl-5-azaspiro-2-4-heptane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84713
Availability details for
Quantity
  • Availability
  • Add to Cart