ADVH49B8290F AChemBlock

methyl 1-ethylindole-4-carboxylate

  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-ethylindole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84958
Availability details for
Quantity
  • Availability
  • Add to Cart