ADVH49B82911 AChemBlock

methyl 1-[2-(N'-hydroxycarbamimidoyl)ethyl]pyrrolidine-2-carboxylate

  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-2-n-hydroxycarbamimidoyl-ethyl-pyrrolidine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84960
Availability details for
Quantity
  • Availability
  • Add to Cart