ADVH49B82916 AChemBlock

methyl 1-(4-bromopyrazol-1-yl)cyclopropanecarboxylate

  • Molecular Weight : 245.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-4-bromopyrazol-1-yl-cyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84965
Availability details for
Quantity
  • Availability
  • Add to Cart