ADVH49B82919 AChemBlock

tert-butyl (6R)-6-(aminomethyl)-5-azaspiro[2.4]heptane-5-carboxylate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6r-6-aminomethyl-5-azaspiro-2-4-heptane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84718
Availability details for
Quantity
  • Availability
  • Add to Cart