ADVH49B8291D AChemBlock

methyl (3S)-1-oxo-2-azaspiro[4.4]nonane-3-carboxylate

  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3s-1-oxo-2-azaspiro-4-4-nonane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84975
Availability details for
Quantity
  • Availability
  • Add to Cart