ADVH49B82922 AChemBlock

methyl (3R)-3-methyl-2,3-dihydro-1H-indole-4-carboxylate hydrochloride

  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3r-3-methyl-2-3-dihydro-1h-indole-4-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84980
Availability details for
Quantity
  • Availability
  • Add to Cart