ADVH49B82923 AChemBlock

methyl (3R)-3-fluoropyrrolidine-3-carboxylate hydrochloride

  • Molecular Weight : 183.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3r-3-fluoropyrrolidine-3-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84981
Availability details for
Quantity
  • Availability
  • Add to Cart