ADVH49B82924 AChemBlock

methyl (2S,4R)-4-[tert-butoxycarbonyl(methyl)amino]pyrrolidine-2-carboxylate

  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2s-4r-4-tert-butoxycarbonyl-methyl-amino-pyrrolidine-2-carboxylate-308292.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84982
Availability details for
Quantity
  • Availability
  • Add to Cart