ADVH49B82925 AChemBlock

methyl (2S,3S)-rel-3-hydroxypyrrolidine-2-carboxylate hydrochloride

  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2s-3s-rel-3-hydroxypyrrolidine-2-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84983
Availability details for
Quantity
  • Availability
  • Add to Cart