ADVH49B82926 AChemBlock

methyl (2S)-2-amino-2-(oxetan-3-yl)acetate

  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2s-2-amino-2-oxetan-3-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84984
Availability details for
Quantity
  • Availability
  • Add to Cart