ADVH49B82927 AChemBlock

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]propanoate

  • Molecular Weight : 395.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2s-2-tert-butoxy-carbonyl-amino-3-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84985
Availability details for
Quantity
  • Availability
  • Add to Cart