ADVH49B82928 AChemBlock

methyl (2R,4S)-4-sulfanylpyrrolidine-2-carboxylate;hydrochloride

  • Molecular Weight : 197.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2r-4s-4-sulfanylpyrrolidine-2-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84986
Availability details for
Quantity
  • Availability
  • Add to Cart