ADVH49B8292F AChemBlock

methyl (1R,3R)-rel-3-(1,1-difluoroethyl)cyclopentanecarboxylate

  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1r-3r-rel-3-1-1-difluoroethyl-cyclopentanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84993
Availability details for
Quantity
  • Availability
  • Add to Cart