ADVH49B82930 AChemBlock

methyl (1R,3R)-3-amino-1-methyl-cyclopentanecarboxylate hydrochloride

  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1r-3r-3-amino-1-methyl-cyclopentanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84994
Availability details for
Quantity
  • Availability
  • Add to Cart