ADVH49B82932 AChemBlock

lithium(1+) (4-fluoropyridin-2-yl)trihydroxyboranuide

  • Molecular Weight : 164.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/lithium-1-4-fluoropyridin-2-yl-trihydroxyboranuide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84996
Availability details for
Quantity
  • Availability
  • Add to Cart