ADVH49B82933 AChemBlock

tert-butyl 3a-methyl-1,2,3,4,6,6a-hexahydropyrrolo[3,4-c]pyrrole-5-carboxylate hydrochloride

  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3a-methyl-1-2-3-4-6-6a-hexahydropyrrolo-3-4-c-pyrrole-5-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84651
Availability details for
Quantity
  • Availability
  • Add to Cart