ADVH49B82935 AChemBlock

tert-butyl 3-{[(4-methylbenzenesulfonyl)oxy]methyl}-4H,5H,6H,7H-[1,2]oxazolo[4,5-c]pyridine-5-carboxylate

  • Molecular Weight : 408.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-4-methylbenzenesulfonyl-oxy-methyl-4h-5h-6h-7h-1-2-oxazolo-4-5-c-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84653
Availability details for
Quantity
  • Availability
  • Add to Cart