ADVH49B82939 AChemBlock

imidazo[1,5-c]pyrimidin-5-amine

  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-5-c-pyrimidin-5-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85002
Availability details for
Quantity
  • Availability
  • Add to Cart