ADVH49B8293E AChemBlock

exo-6-methoxy-3-azabicyclo[3.1.0]hexane hydrochloride

  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-6-methoxy-3-azabicyclo-3-1-0-hexane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85006
Availability details for
Quantity
  • Availability
  • Add to Cart