ADVH49B8293F AChemBlock

exo-6-ethynyl-3-oxabicyclo[3.1.0]hexane

  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-6-ethynyl-3-oxabicyclo-3-1-0-hexane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85007
Availability details for
Quantity
  • Availability
  • Add to Cart