ADVH49B82941 AChemBlock

exo-3-benzyl-3-azabicyclo[3.1.1]heptan-6-amine dihydrochloride

  • Molecular Weight : 275.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-3-benzyl-3-azabicyclo-3-1-1-heptan-6-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85009
Availability details for
Quantity
  • Availability
  • Add to Cart