ADVH49B82942 AChemBlock

exo-(1R,2R,4S)-bicyclo[2.2.1]heptane-2-carboxylic acid

  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-1r-2r-4s-bicyclo-2-2-1-heptane-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85011
Availability details for
Quantity
  • Availability
  • Add to Cart