ADVH49B82943 AChemBlock

tert-butyl (5S)-3,3-difluoro-5-(3-oxobutanoyl)piperidine-1-carboxylate

  • Molecular Weight : 305.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5s-3-3-difluoro-5-3-oxobutanoyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84725
Availability details for
Quantity
  • Availability
  • Add to Cart