ADVH49B82945 AChemBlock

ethyl trans-4-(difluoromethyl)pyrrolidine-3-carboxylate

  • Molecular Weight : 193.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-trans-4-difluoromethyl-pyrrolidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85014
Availability details for
Quantity
  • Availability
  • Add to Cart