ADVH49B82946 AChemBlock

tert-butyl (5S)-2,7-diazaspiro[4.4]nonane-2-carboxylate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5s-2-7-diazaspiro-4-4-nonane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84726
Availability details for
Quantity
  • Availability
  • Add to Cart