ADVH49B82947 AChemBlock

tert-butyl 3-{[(4-methylbenzenesulfonyl)oxy]methyl}-4H,5H,6H,7H,7aH-pyrazolo[3,4-c]pyridine-6-carboxylate

  • Molecular Weight : 407.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-4-methylbenzenesulfonyl-oxy-methyl-4h-5h-6h-7h-7ah-pyrazolo-3-4-c-pyridine-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84655
Availability details for
Quantity
  • Availability
  • Add to Cart